4-Methoxybenzonitrile
Glass bottle for solids, 25 g
Catalog # MOLEM37711531
Supplier: Molekula
Jump to:
Product Details & Documents
Synonyms: 4-Cyanoanisol, Anisonitrile, p-Cyanoanisol, p-Methoxybenzonitrile
- CAS Number:874-90-8
- Formula:C₈H₇NO
- MDL Number:MFCD00001818
- Molecular Weight:133.15 g/mol
- Boiling Pt:256 °C (1013 hPa)
- Melting Pt:57…60 °C
- Storage Temperature:Ambient
- EINECS:212-871-6
- UN:3439
- ADR:6.1,III
Specifications
Specifications
- Pack type:Glass bottle for solids
- Pk:25 g



