Order Entry
Export
Orders LinkContactUsLinkComponent
4-Methoxybenzonitrile

4-Methoxybenzonitrile

Irritant
Supplier:   CHEMPUR
4-Methoxybenzonitrile
Supplier:  CHEMPUR
Certificates
4-Methoxybenzonitrile
4-Methoxybenzonitrile

2 Options Available Below

Product Details & Documents
Synonyms:  4-Cyanoanisol, Anisonitrile, p-Cyanoanisol, p-Methoxybenzonitrile
  • CAS Number:874-90-8
  • Formula:C₈H₇NO
  • MDL Number:MFCD00001818
  • Molecular Weight:133.15 g/mol
  • Boiling Pt:256 °C (1013 hPa)
  • Melting Pt:57…60 °C
  • Storage Temperature:Ambient
  • EINECS:212-871-6
  • UN:3439
  • ADR:6.1,III
Product Family Options

Product Information

  • Pack type
    Pk
    Availability
    Price
  • Glass bottle for solids
    250 mg
    Log in for availability
    Log in for availability
    1 * 250 mg
    1 * 250 mg

  • Glass bottle for solids
    1 g
    Log in for availability
    Log in for availability
    1 * 1 g
    1 * 1 g

This product has been blocked by your organization. Please contact your purchasing department for more information