2-Nitrobenzonitrile
Glass bottle for solids, 5 g
MOLE19854774-5G
: Molekula
: o-Nitrobenzonitrile
- :612-24-8
- :C₇H₄N₂O₂
- :MFCD00007044
- :148.12 g/mol
- :304 °C (1013 hPa)
- :107…109 °C
- :Ambient
- :210-301-0
- :3439
- :6.1,II
- Pack type:Glass bottle for solids
- Pk:5 g