Order Entry
Export
Orders LinkContactUsLinkComponent
 

2-Nitrobenzonitrile 98%

Glass bottle, 10 g
Catalog # ACRO128470100
Toxic
2-Nitrobenzonitrile 98%
Catalog # ACRO128470100
CAS Number:  612-24-8
2-Nitrobenzonitrile 98%
2-Nitrobenzonitrile 98%

Glass bottle, 10 g

CAS Number:  612-24-8
Price
37,60 €
Log in for availability
1 * 10 g
Product Details & Documents
Synonyms:  o-Nitrobenzonitrile
  • CAS Number:612-24-8
  • Formula:C₇H₄N₂O₂
  • MDL Number:MFCD00007044
  • Molecular Weight:148.12 g/mol
  • Boiling Pt:165 °C (16 mmHg)
  • Melting Pt:110.8…111.8 °C
  • Storage Temperature:Ambient
  • EINECS:210-301-0
  • UN:3439
  • ADR:6.1,II

 

Specifications

Specification Test Results

  • Appearance:
    Yellow Crystalline powder
  • Infrared spectrum:
    Conforms
  • Melting point:
    107 to 112 °C
  • GC:
    ≥97.5 %

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    10 g

Specifications