Order Entry
Export
Orders LinkContactUsLinkComponent
3,5-Dihydroxybenzaldehyde ≥98%

3,5-Dihydroxybenzaldehyde ≥98%

3,5-Dihydroxybenzaldehyde ≥98%
Certificates
3,5-Dihydroxybenzaldehyde ≥98%
3,5-Dihydroxybenzaldehyde ≥98%

3 Options Available Below

Product Details & Documents

Purity: 98% CAS No.: 26153-38-8 Molecular Formula: C7H6O3 Formula Weight: 138.12 Physical Form: Powder Sensitivity: Air Sensitive Beilstein Reference: 1930147 MDL No.: MFCD00016611

  • CAS Number:26153-38-8
  • Formula:C₇H₆O₃
  • MDL Number:MFCD00016611
  • Molecular Weight:138.12 g/mol
  • Melting Pt:158…161 °C
  • Storage Temperature:Ambient
  • EINECS:247-479-4
Product Family Options

Product Information