3,4-Dihydroxybenzaldehyde, Sigma-Aldrich®
Supplier: Merck
Jump to:
About this item
Synonyms: Protocatechuic aldehyde, Protocatechualdehyde
- CAS Number:139-85-5
- Formula:C₇H₆O₃
- MDL Number:MFCD00003370
- Molecular Weight:138.12 g/mol
- Merck Index:12,08079
- Boiling Pt:295 °C (1013 hPa)
- Melting Pt:150…155 °C
- Density:1.505 g/cm³ (20 °C)
- :M-Clarity™ quality level = MQ100
- Storage Temperature:Ambient
- EINECS:205-377-7
2 Options Available Below
Product Details & Documents
Synonyms: Protocatechuic aldehyde, Protocatechualdehyde
- CAS Number:139-85-5
- Formula:C₇H₆O₃
- MDL Number:MFCD00003370
- Molecular Weight:138.12 g/mol
- Merck Index:12,08079
- Boiling Pt:295 °C (1013 hPa)
- Melting Pt:150…155 °C
- Density:1.505 g/cm³ (20 °C)
- :M-Clarity™ quality level = MQ100
- Storage Temperature:Ambient
- EINECS:205-377-7



