3,4-Dihydroxybenzaldehyde 97%
: Protocatechuic aldehyde, Protocatechualdehyde
- :139-85-5
- :C₇H₆O₃
- :MFCD00003370
- :138.12 g/mol
- :12,08079
- :295 °C (1013 hPa)
- :152…157 °C
- :1,505 g/cm³ (20 °C)
- :Ambient
- :205-377-7
: Protocatechuic aldehyde, Protocatechualdehyde
- :139-85-5
- :C₇H₆O₃
- :MFCD00003370
- :138.12 g/mol
- :12,08079
- :295 °C (1013 hPa)
- :152…157 °C
- :1,505 g/cm³ (20 °C)
- :Ambient
- :205-377-7
Documents
ACRO11485
SDS
- Appearance:Beige to grey-brown Crystalline powder
- Infrared spectrum:Conforms
- Melting point:152 to 157 °C
- HPLC:≥96.0 %
- Copper (Cu):≤50 ppm
- Iron (Fe):≤100 ppm
- Lead (Pb):≤50 ppm
Product Family Options
- Pack typePk



