2,5-Dihydroxybenzaldehyde ≥98%
Supplier: Thermo Fisher Scientific
Jump to:
About this item
Purity: 98+% CAS No.: 1194-98-5 Molecular Formula: C7H6O3 Formula Weight: 138.12 Melting Point: 97-101° Beilstein Reference: 1363961
- CAS Number:1194-98-5
- Formula:C₇H₆O₃
- MDL Number:MFCD00003333
- Molecular Weight:138.12 g/mol
- Boiling Pt:60…65 °C (12 mmHg)
- Melting Pt:97…101 °C
- Density:0.859 g/cm³ (20 °C)
- Storage Temperature:Ambient
- EINECS:214-789-6
3 Options Available Below
Product Details & Documents
Purity: 98+% CAS No.: 1194-98-5 Molecular Formula: C7H6O3 Formula Weight: 138.12 Melting Point: 97-101° Beilstein Reference: 1363961
- CAS Number:1194-98-5
- Formula:C₇H₆O₃
- MDL Number:MFCD00003333
- Molecular Weight:138.12 g/mol
- Boiling Pt:60…65 °C (12 mmHg)
- Melting Pt:97…101 °C
- Density:0.859 g/cm³ (20 °C)
- Storage Temperature:Ambient
- EINECS:214-789-6



