Order Entry
Export
Orders LinkContactUsLinkComponent
2,5-Dihydroxybenzaldehyde 99%

2,5-Dihydroxybenzaldehyde 99%

2,5-Dihydroxybenzaldehyde 99%
 
2,5-Dihydroxybenzaldehyde 99%
2,5-Dihydroxybenzaldehyde 99%

 

 
  •  :1194-98-5
  •  :C₇H₆O₃
  •  :MFCD00003333
  •  :138.12 g/mol
  •  :60 - 65 °C (12 -rr)
  •  :98…102 °C
  •  :0,859 g/cm³ (20 °C)
  •  :Ambient
  •  :214-789-6

Documents

 

 

  • Appearance:
    Yellow to khaki Crystalline powder
  • Infrared spectrum:
    Conforms
  • Melting point:
    98 to 102 °C
  • GC:
    ≥98.5 %
  • Copper (Cu):
    ≤50 ppm
  • Iron (Fe):
    ≤100 ppm
  • Lead (Pb):
    ≤50 ppm
Product Family Options

 

  • Pack type
    Pk
     
     
  • Glass bottle for solids
    5 g
       
       
    1 * 5 g
    1 * 5 g - 72,50 €
    72,50 €
    1 * 5 g
       72,50 €

  • Glass bottle
    25 g
       
       
    1 * 25 g
    1 * 25 g - 255,00 €
    255,00 €
    1 * 25 g
       255,00 €