2,4-Dihydroxybenzaldehyde 98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:95-01-2
:
Formula:C₇H₆O₃
MDL Number:MFCD00011686
:138.12 g/mol
Merck Index:12,08324
Boiling Pt:220…228 °C (22 mmHg)
Melting Pt:133…138 °C
Density:1,448 g/cm³ (-93 °C)
EINECS:202-383-1
:GHS07
Storage Temperature:Ambient
Specification Test Results
- Appearance:Light beige to pink-orange Powder
- Infrared spectrum:Conforms
- Melting point:134 to 140 °C
- GC:≥97.5 %