2,3-Dihydroxybenzaldehyde
Glass bottle for solids, 25 g
Catalog # MOLE21713550-25G
Supplier: Molekula
Jump to:
Product Details & Documents
- CAS Number:24677-78-9
- Formula:C₇H₆O₃
- MDL Number:MFCD00003324
- Molecular Weight:138.12 g/mol
- Boiling Pt:235 °C (1013 hPa)
- Melting Pt:103…105 °C
- Storage Temperature:Ambient
- EINECS:246-398-1
Specifications
Specifications
- Pack type:Glass bottle for solids
- Pk:25 g



