2,3-Dihydroxybenzaldehyde ≥97%
Supplier: Thermo Fisher Scientific
Jump to:
About this item
Purity: 97% CAS No.: 24677-78-9 Molecular Formula: C7H6O3 Formula Weight: 138.12 Melting Point: 106-109° Beilstein Reference: 2041781
- CAS Number:24677-78-9
- Formula:C₇H₆O₃
- MDL Number:MFCD00003324
- Molecular Weight:138.12 g/mol
- Boiling Pt:235 °C (1013 hPa)
- Melting Pt:106…109 °C
- Storage Temperature:Ambient
- EINECS:246-398-1
3 Options Available Below
Product Details & Documents
Purity: 97% CAS No.: 24677-78-9 Molecular Formula: C7H6O3 Formula Weight: 138.12 Melting Point: 106-109° Beilstein Reference: 2041781
- CAS Number:24677-78-9
- Formula:C₇H₆O₃
- MDL Number:MFCD00003324
- Molecular Weight:138.12 g/mol
- Boiling Pt:235 °C (1013 hPa)
- Melting Pt:106…109 °C
- Storage Temperature:Ambient
- EINECS:246-398-1



