3,5-Dimethoxybenzaldehyde
Glass bottle for solids, 25 g
Catalog # MOLEM11112649
Supplier: Molekula
Jump to:
Product Details & Documents
- CAS Number:7311-34-4
- Formula:C₉H₁₀O₃
- MDL Number:MFCD00003366
- Molecular Weight:166.18 g/mol
- Boiling Pt:277 °C (1013 hPa)
- Melting Pt:48 °C
- Storage Temperature:Ambient
- EINECS:230-772-6
Specifications
Specifications
- Pack type:Glass bottle for solids
- Pk:25 g



