2,6-Dimethoxybenzaldehyde
Supplier: Thermo Scientific
Jump to:
About this item
- CAS Number:3392-97-0
- Formula:C₉H₁₀O₃
- MDL Number:MFCD00010862
- Molecular Weight:166.18 g/mol
- Boiling Pt:285 °C (1013 hPa)
- Melting Pt:95…96 °C
- Storage Temperature:Refrigerator
2 Options Available Below
Product Details & Documents
- CAS Number:3392-97-0
- Formula:C₉H₁₀O₃
- MDL Number:MFCD00010862
- Molecular Weight:166.18 g/mol
- Boiling Pt:285 °C (1013 hPa)
- Melting Pt:95…96 °C
- Storage Temperature:Refrigerator
Product Family Options
Product Information
- Pack typePkAvailabilityPrice



