2,6-Dimethoxybenzaldehyde 99%
Glass bottle for solids, 1 g
Supplier: Thermo Fisher Scientific
2,6-Dimethoxybenzaldehyde 99%
Catalog # ACRO305790010
Supplier: Thermo Fisher Scientific
CAS Number: 3392-97-0
Jump to:
Product Details & Documents
- CAS Number:3392-97-0
- Formula:C₉H₁₀O₃
- MDL Number:MFCD00010862
- Molecular Weight:166.18 g/mol
- Boiling Pt:285 °C (1013 hPa)
- Melting Pt:95…98 °C
- Storage Temperature:Refrigerator
Documents
ACRO30579
SDS
Specifications
Specification Test Results
- Appearance:Yellow to beige Crystalline powder
- Infrared spectrum:Conforms
- Melting point:95 to 98 °C
- GC:≥98.5 %
Specifications
- Pack type:Glass bottle for solids
- Pk:1 g



