Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
2,5-Dimethoxybenzaldehyde
2,5-Dimethoxybenzaldehyde ≥98%
2,5-Dimethoxybenzaldehyde ≥98%
:
Thermo Fisher Scientific
2,5-Dimethoxybenzaldehyde ≥98%
:
Thermo Fisher Scientific
:
93-02-7
:
:
C₉H₁₀O₃
:
MFCD00003314
:
166.18 g/mol
:
146 °C (10 mmHg)
:
47…51 °C
:
>110 °C (230 °F)
:
Ambient
:
202-211-5
:
GHS07
msds_ALFAA19928
Organic Building Blocks and Intermediates
Thermo Scientific Acroseal 25 Years