Order Entry
Export
ContactUsLinkComponent
2,4-Dimethoxybenzaldehyde 98%
Irritant
2,4-Dimethoxybenzaldehyde 98%
Certificates
undefined
Irritant
2,4-Dimethoxybenzaldehyde 98%

Specification Test Results

  • Appearance:
    White to light brown Crystalline powder
  • Infrared spectrum:
    Conforms
  • Melting point:
    68 to 72 °C
  • GC:
    ≥97.5 %

Specification Test Results

Related Information