
4-Hydroxy-2-methoxybenzaldehyde, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:18278-34-7
:
Formula:C₈H₈O₃
MDL Number:MFCD00051964
:152.15 g/mol
Boiling Pt:273 °C (1013 hPa)
Melting Pt:156…158 °C
:GHS07
Storage Temperature:Ambient