D-(+)-Trehalose dihydrate Ph. Eur., USP, NF
Catalog # VWRS6073.9010
Supplier: VWR Collection
CAS Number:
Specifications
- Pk:10 kg
- Seq:4158
Specifications
About this item
:
:378.33 g/mol
Melting Pt:97…99 °C
Storage Temperature:Ambient
EINECS:202-739-6
CAS Number:6138-23-4
Formula:C₁₂H₂₂O₁₁·2H₂O
MDL Number:MFCD00071594