Order Entry
Export
Orders LinkContactUsLinkComponent
4-Hydroxy-2-methoxybenzaldehyde 98%

4-Hydroxy-2-methoxybenzaldehyde 98%

Irritant
4-Hydroxy-2-methoxybenzaldehyde 98%
Certificates
4-Hydroxy-2-methoxybenzaldehyde 98%

3 Options Available Below

Product Details & Documents
  • CAS Number:18278-34-7
  • Formula:C₈H₈O₃
  • MDL Number:MFCD00051964
  • Molecular Weight:152.15 g/mol
  • Boiling Pt:273 °C (1013 hPa)
  • Melting Pt:153…158 °C
  • Storage Temperature:Ambient

Documents

Specifications for Product Family

Specification Test Results

  • Appearance:
    Off-white to light brown Powder
  • Infrared spectrum:
    Conforms
  • Melting point:
    156 to 161 °C
  • HPLC:
    ≥97.5 %
Product Family Options

Product Information

  • Pack type
    Pk
    Availability
    Price
  • Glass bottle for solids
    1 g
    Log in for availability
    Log in for availability
    1 * 1 g
    1 * 1 g - 36,60 €
    36,60 €
    1 * 1 g
       36,60 €

  • Glass bottle for solids
    5 g
    Log in for availability
    Log in for availability
    1 * 5 g
    1 * 5 g - 118,00 €
    118,00 €
    1 * 5 g
       118,00 €

  • Glass bottle
    25 g
    Log in for availability
    Log in for availability
    1 * 25 g
    1 * 25 g - 454,00 €
    454,00 €
    1 * 25 g
       454,00 €