Order Entry
Export
ContactUsLinkComponent
3-Hydroxy-p-anisaldehyde
3-Hydroxy-p-anisaldehyde
Catalog # MOLE26504291-100G
Supplier:  Molekula
CAS Number:  
3-Hydroxy-p-anisaldehyde
Catalog # MOLE26504291-100G
Supplier:  Molekula
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    100 g
  • Seq:
    3401

Specifications

About this item

 :
 :152.15 g/mol
Boiling Pt:308 °C (1013 hPa)
Melting Pt:68…70 °C
Density:1.401 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:210-694-9
CAS Number:621-59-0
Formula:C₈H₈O₃
MDL Number:MFCD00003369