Order Entry
Export
ContactUsLinkComponent
3-Hydroxy-p-anisaldehyde 98%
Irritant
3-Hydroxy-p-anisaldehyde 98%
Certificates
undefined
Irritant
3-Hydroxy-p-anisaldehyde 98%

Specification Test Results

  • Appearance:
    Off-white to grey to light brown Powder
  • Infrared spectrum:
    Conforms
  • Melting point:
    112 to 116 °C
  • GC:
    ≥97.5 %

Specification Test Results

Related Information