3-Ethoxybenzaldehyde ≥97%
Supplier: Thermo Fisher Scientific
Jump to:
About this item
Purity: 97% CAS No.: 22924-15-8 Molecular Formula: C9H10O2 Formula Weight: 150.18 Boiling Point: 132-134/15mm MDL No.: MFCD00016606
- CAS Number:22924-15-8
- Formula:C₉H₁₀O₂
- MDL Number:MFCD00016606
- Molecular Weight:150.18 g/mol
- Boiling Pt:132…134 °C (15 mmHg)
- Melting Pt:21 °C
- Density:1.07
- Storage Temperature:Ambient
- EINECS:245-333-4
2 Options Available Below
Product Details & Documents
Purity: 97% CAS No.: 22924-15-8 Molecular Formula: C9H10O2 Formula Weight: 150.18 Boiling Point: 132-134/15mm MDL No.: MFCD00016606
- CAS Number:22924-15-8
- Formula:C₉H₁₀O₂
- MDL Number:MFCD00016606
- Molecular Weight:150.18 g/mol
- Boiling Pt:132…134 °C (15 mmHg)
- Melting Pt:21 °C
- Density:1.07
- Storage Temperature:Ambient
- EINECS:245-333-4
Documents
Organic Building Blocks and Intermediates



