
4-Hydroxy-3-nitrobenzaldehyde ≥98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:3011-34-5
:
Formula:C₇H₅NO₄
MDL Number:MFCD00007117
:167.12 g/mol
Melting Pt:140…144 °C
EINECS:221-141-6
:GHS07
Storage Temperature:Ambient