Order Entry
Export
ContactUsLinkComponent
5-Nitrovanillin
5-Nitrovanillin
Catalog # MOLE23063193-25G
Supplier:  Molekula
CAS Number:  
5-Nitrovanillin
Catalog # MOLE23063193-25G
Supplier:  Molekula
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    25 g
  • Seq:
    3308
  • Environmentally Preferable:

Specifications

About this item

Synonyms: 4-Hydroxy-3-methoxy-5-nitrobenzaldehyde
 :
 :197.15 g/mol
Melting Pt:175…178 °C
Storage Temperature:Ambient
EINECS:229-633-2
CAS Number:6635-20-7
Formula:C₈H₇NO₅
MDL Number:MFCD00007118