4-(Dimethylamino)benzaldehyde
Glass bottle for solids, 500 g
Catalog # MOLE52742798-500G
Supplier: Molekula
Jump to:
Product Details & Documents
Synonyms: Ehrlich's Reagent, 4-(N,N-Dimethylamino)benzaldehyde, p-Dimethylamino-Benzaldehyde, p-Dimethylaminobenzaldehyde, p-(Dimethylamino)benzaldehyde
- CAS Number:100-10-7
- Formula:(CH₃)₂NC₆H₄CHO
- MDL Number:MFCD00003381
- Molecular Weight:149.19 g/mol
- Merck Index:13,03257
- Boiling Pt:176…177 °C (1013 hPa)
- Melting Pt:72 °C
- Density:1.0254 g/cm³ (99.9 °C)
- Flash Pt:>100 °C
- Storage Temperature:Ambient
- EINECS:202-819-0
Specifications
Specifications
- Pack type:Glass bottle for solids
- Pk:500 g



