Orders LinkContactUsLinkComponent
4-(Dimethylamino)benzaldehyde 98%
4-(Dimethylamino)benzaldehyde 98%
4-(Dimethylamino)benzaldehyde 98%
4-(Dimethylamino)benzaldehyde 98%
Purity: 98% CAS No.: 100-10-7 Molecular Formula: C9H11NO Formula Weight: 149.19 Flash Point: 164°(327°F) Odour: Strong Boiling Point: 176-177°/17mm Beilstein Reference: 606802 Merck Reference: 14,3230 MDL No.: MFCD00003381
 :  Ehrlich's Reagent, 4-(N,N-Dimethylamino)benzaldehyde, p-Dimethylamino-Benzaldehyde, p-Dimethylaminobenzaldehyde, p-(Dimethylamino)benzaldehyde
  •  :100-10-7
  •  :(CH₃)₂NC₆H₄CHO
  •  :MFCD00003381
  •  :149.19 g/mol
  •  :14,3230
  •  :176…177 °C (17 mmHg)
  •  :73…75 °C
  •  :1.0254 g/cm³ (99.9 °C)
  •  :164 °C (327 °F)
  •  :Ambient
  •  :202-819-0
Product Family Options
  • Pack type
    Pk
  • Glass bottle for solids
    100 g
    1 * 100 g
    1 * 100 g - 38,80 €
    38,80 €
    1 * 100 g
       38,80 €
  • Glass bottle for solids
    500 g
    1 * 500 g
    1 * 500 g - 139,00 €
    139,00 €
    1 * 500 g
       139,00 €
  • Glass bottle for solids
    2.500 g
    1 * 2.500 g
    1 * 2.500 g - 480,00 €
    480,00 €
    1 * 2.500 g
       480,00 €