
2,6-Dibromoaniline 97%
Catalog # ACRO112700100
Supplier: Thermo Fisher Scientific
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:10 g
- Seq:3236
Specifications
Specification Test Results
- Appearance:White to yellow to brown Crystalline powder or solid
- Infrared spectrum:Conforms
- Melting point:79 to 84 °C
- GC:≥96.0 %
Specification Test Results
About this item
:
:250.92 g/mol
Boiling Pt:262…264 °C (760 mmHg)
Melting Pt:79…84 °C
Storage Temperature:Ambient
EINECS:210-158-4
ADR:6.1,III
:GHS07
CAS Number:608-30-0
Formula:C₆H₅Br₂N
MDL Number:MFCD00007638