

2,5-Dibromoaniline 97%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:3638-73-1
:
Formula:C₆H₅Br₂N
MDL Number:MFCD00007636
:250.92 g/mol
Boiling Pt:281 °C (1013 hPa)
Melting Pt:52…56 °C
Flash Pt:>110 °C (230 °F)
Storage Temperature:Ambient
EINECS:222-865-5
UN:2811
ADR:6.1,III
:GHS06
:GHS07