1-Bromo-4-(trifluoromethoxy)benzene
Supplier: Thermo Scientific
Certificates
About this item
CAS Number:407-14-7
:
Formula:C₇H₄BrF₃O
MDL Number:MFCD00040834
:241.01 g/mol
Boiling Pt:153 °C (1013 hPa)
Density:1.62 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:206-979-2
UN:3334
ADR:9,