2-Hydroxy-5-methylpyridine, Sigma-Aldrich®
Catalog # SIAL593427-1G
Supplier: Merck
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:109.13 g/mol
:M-Clarity™ quality level = MQ100
EINECS:213-713-9
CAS Number:1003-68-5
:91914-06-6
Formula:C₆H₇NO
MDL Number:MFCD03092888