Order Entry
Export
ContactUsLinkComponent
1,8-Diiodohexadecafluorooctane, Sigma-Aldrich®
1,8-Diiodohexadecafluorooctane, Sigma-Aldrich®
Catalog # SIAL484997-1G
Supplier:  Merck
CAS Number:  
1,8-Diiodohexadecafluorooctane, Sigma-Aldrich®
Catalog # SIAL484997-1G
Supplier:  Merck
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :653.87 g/mol
 :M-Clarity™ quality level = MQ100
CAS Number:335-70-6
Formula:C₈F₁₆I₂
MDL Number:MFCD00236093