2-Chloro-4,6-dimethoxy-1,3,5-triazine, Sigma-Aldrich®
Catalog # SIAL375217-5G
Supplier: Merck
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:5 g
- Seq:3173
Specifications
About this item
:
:175.57 g/mol
:M-Clarity™ quality level = MQ200
EINECS:221-541-0
CAS Number:3140-73-6
Formula:C₅H₆ClN₃O₂
MDL Number:MFCD00075607