Order Entry
Export
ContactUsLinkComponent
 
(1S,2S)-(-)-1,2-Diphenylethylenediamine, Sigma-Aldrich®
Catalog # SIAL364002-500MG
Supplier:  Merck
CAS Number:  
(1S,2S)-(-)-1,2-Diphenylethylenediamine, Sigma-Aldrich®
Catalog # SIAL364002-500MG
Supplier:  Merck
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    500 mg
  • Seq:
    2725

Specifications

About this item

 :
 :M-Clarity™ quality level = MQ200
 :212.29 g/mol
CAS Number:29841-69-8
Formula:C₁₄H₁₆N₂
MDL Number:MFCD00082751