1,3-Dibenzoylpropane, Sigma-Aldrich®
Catalog # SIAL362069-5G
Supplier: Merck
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:5 g
- Seq:3173
Specifications
About this item
:
:252.31 g/mol
CAS Number:6263-83-8
Formula:C₁₇H₁₆O₂
MDL Number:MFCD00051401