Order Entry
Export
ContactUsLinkComponent
3,4-Dichlorophenylhydrazinium chloride, Sigma-Aldrich®
3,4-Dichlorophenylhydrazinium chloride, Sigma-Aldrich®
Catalog # SIAL152811-25G
Supplier:  Merck
CAS Number:  
3,4-Dichlorophenylhydrazinium chloride, Sigma-Aldrich®
Catalog # SIAL152811-25G
Supplier:  Merck
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    25 g
  • Seq:
    3308

Specifications

About this item

 :
 :213.49 g/mol
 :M-Clarity™ quality level = MQ100
EINECS:243-278-0
CAS Number:19763-90-7
Formula:C₆H₇Cl₃N₂
MDL Number:MFCD00012937