Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
3-Bromo-4,5-dimethoxybenzaldehyde
3-Bromo-4,5-dimethoxybenzaldehyde, Sigma-Aldrich®
3-Bromo-4,5-dimethoxybenzaldehyde, Sigma-Aldrich®
3-Bromo-4,5-dimethoxybenzaldehyde, Sigma-Aldrich®
SIAL130613-5G
:
Merck
:
3-Bromo-4,5-dimethoxybenzaldehyde, Sigma-Aldrich®
SIAL130613-5G
:
Merck
:
Pack type:
Glass bottle for solids
Pk:
5 g
Seq:
3173
:
:
245.07 g/mol
:
M-Clarity™ quality level = MQ100
:
230-113-2
:
GHS07
:
6948-30-7
:
C₉H₉BrO₃
:
MFCD00003346