Order Entry
Export
Orders LinkContactUsLinkComponent
Methyl-3,5-dimethoxybenzoate

Methyl-3,5-dimethoxybenzoate

Glass bottle for solids, 100 g
Catalog # MOLE82959542-100G
Supplier:   Molekula
Methyl-3,5-dimethoxybenzoate
Catalog # MOLE82959542-100G
Supplier:  Molekula
CAS Number:  2150-37-0
Methyl-3,5-dimethoxybenzoate

Glass bottle for solids, 100 g

CAS Number:  2150-37-0
Price
Inquire For Price
Log in for availability
1 * 100 g
Product Details & Documents
Synonyms:  Methyl 3,5-dimethoxybenzoate, 3,5-Dimethoxybenzoic acid methyl ester
  • CAS Number:2150-37-0
  • Formula:C₁₀H₁₂O₄
  • MDL Number:MFCD00008432
  • Molecular Weight:196.2 g/mol
  • EINECS:218-423-6
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    100 g

Specifications