2-Fluorophenylhydrazinium chloride
Glass bottle for solids, 5 g
Catalog # MOLEM62132853
Supplier: Molekula
Jump to:
Product Details & Documents
Synonyms: (2-Fluorophenyl)hydrazine hydrochloride, (2-Fluorphenyl)hydrazin HCl, 1-(2-Fluorophenyl)hydrazine hydrochloride, 2-Fluorophenylhydrazine hydrochloride
- CAS Number:2924-15-4
- Formula:C₆H₈ClFN₂
- MDL Number:MFCD00012927
- Molecular Weight:162.59 g/mol
- EINECS:220-885-9
Specifications
Specifications
- Pack type:Glass bottle for solids
- Pk:5 g
- Cat. No.:MOLEM62132853



