(1R,2R)-(+)-1,2-Diphenylethylenediamine
Catalog # MOLE59906939-1G
Supplier: Molekula
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:212.29 g/mol
CAS Number:35132-20-8
Formula:C₁₄H₁₆N₂
MDL Number:MFCD00082769