Order Entry
Export
ContactUsLinkComponent
(1S,2S)-(-)-1,2-Diphenylethylenediamine
(1S,2S)-(-)-1,2-Diphenylethylenediamine
Catalog # MOLEM18630987
Supplier:  Molekula
CAS Number:  
(1S,2S)-(-)-1,2-Diphenylethylenediamine
Catalog # MOLEM18630987
Supplier:  Molekula
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :212.29 g/mol
CAS Number:29841-69-8
Formula:C₁₄H₁₆N₂
MDL Number:MFCD00082751