Organic reference standard, Trichloroethylene 100 µg/ml in methanol, Dr Ehrenstorfer
Catalog # EHERXA17739300ME
Supplier: EHRENSTORFER
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:1 mL
- CAS No.:79-01-6
- Description:Trichloroethylene 100 µg/ml in methanol
- Formula:C₂HCl₃
- Description:Trichloroethylene