Order Entry
Export
ContactUsLinkComponent
Pesticide reference standard, δ-BHC 100 µg/ml in cyclohexane, Dr Ehrenstorfer
Pesticide reference standard, δ-BHC 100 µg/ml in cyclohexane, Dr Ehrenstorfer
Catalog # EHERXA14074000CY
Supplier:  EHRENSTORFER
CAS Number:  
Pesticide reference standard, δ-BHC 100 µg/ml in cyclohexane, Dr Ehrenstorfer
Catalog # EHERXA14074000CY
Supplier:  EHRENSTORFER
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    1 mL
  • CAS No.:
    319-86-8
  • Description:
    δ-BHC 100 µg/ml in cyclohexane
  • Formula:
    C₆H₆Cl₆
  • Description:
    δ-BHC

Specifications

About this item