Organic reference standard, (±)-Epichlorhydrin 100 µg/ml in cyclohexane, Dr Ehrenstorfer
Catalog # EHERXA13175000CY
Supplier: EHRENSTORFER
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:1 mL
- CAS No.:106-89-8
- Description:(±)-Epichlorhydrin 100 µg/ml in cyclohexane
- Formula:C₃H₅ClO
- Description:(±)-Epichlorhydrin