Organic reference standard, 1,4-Diethylbenzene 100 µg/ml in methanol, Dr Ehrenstorfer
Catalog # EHERXA12605600ME
Supplier: EHRENSTORFER
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:1 mL
- CAS No.:105-05-5
- Description:1,4-Diethylbenzene 100 µg/ml in methanol
- Formula:C₁₀H₁₄
- Description:1,4-Diethylbenzene