Organic reference standard, 1,2-Diethylbenzene 100 µg/ml in methanol, Dr Ehrenstorfer
Catalog # EHERXA12605400ME
Supplier: EHRENSTORFER
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:1 mL
- CAS No.:135-01-3
- Description:1,2-Diethylbenzene 100 µg/ml in methanol
- Formula:C₁₀H₁₄
- Description:1,2-Diethylbenzene