Organic reference standard, 1,1-Dichloropropane 100 µg/ml in methanol, Dr Ehrenstorfer
Catalog # EHERXA12479900ME
Supplier: EHRENSTORFER
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:1 mL
- CAS No.:78-99-9
- Description:1,1-Dichloropropane 100 µg/ml in methanol
- Formula:C₃H₆Cl₂
- Description:1,1-Dichloropropane