Organic reference standard, 1,2-Dibromo-1,1-dichloroethane 100 µg/ml in methanol, Dr Ehrenstorfer
Catalog # EHERXA12236500ME
Supplier: EHRENSTORFER
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:1 mL
- CAS No.:75-81-0
- Description:1,2-Dibromo-1,1-dichloroethane 100 µg/ml in methanol
- Formula:C₂H₂Br₂Cl₂
- Description:1,2-Dibromo-1,1-dichloroethane