Order Entry
Export
ContactUsLinkComponent
 
Organic reference standard, o-Xylene 10 µg/ml in methanol, Dr Ehrenstorfer
  EHERL17945000ME
 :  
Organic reference standard, o-Xylene 10 µg/ml in methanol, Dr Ehrenstorfer
  EHERL17945000ME
 :  

 

  • Pack type:
    Glass bottle
  • Pk:
    10 mL
  • CAS No.:
    95-47-6
  • Description:
    o-Xylene 10 µg/ml in methanol
  • Formula:
    C₈H₁₀
  • Description:
    o-Xylene