Order Entry
Export
ContactUsLinkComponent
Pesticide reference standard, 1,3,5-Trichlorobenzene 10 µg/ml in cyclohexane, Dr Ehrenstorfer
Pesticide reference standard, 1,3,5-Trichlorobenzene 10 µg/ml in cyclohexane, Dr Ehrenstorfer
Catalog # EHERL17723500CY
Supplier:  EHRENSTORFER
CAS Number:  
Pesticide reference standard, 1,3,5-Trichlorobenzene 10 µg/ml in cyclohexane, Dr Ehrenstorfer
Catalog # EHERL17723500CY
Supplier:  EHRENSTORFER
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    10 mL
  • CAS No.:
    108-70-3
  • Description:
    1,3,5-Trichlorobenzene 10 µg/ml in cyclohexane
  • Formula:
    C₆H₃Cl₃
  • Description:
    1,3,5-Trichlorobenzene

Specifications

About this item